missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ FE150 Benchtop pH Meter

Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price.

Brand:  Fisherbrand™ S35923

301.83 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 17731739

  • £406.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet FE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.

  • One to five point pH calibration
  • Automatic buffer recognition with two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Meter, Temperature Probe and Stand
±0.2 ±199.9 ±2 Beyond ±200
0 to 14
±1999 mV
USA: pH 1.68, 4.01, 7.00, 10.01, 12.45; NIST: pH 1.68, 4.01, 6.86, 9.18, 12.45
0.1°C
Offset in 0.1°C Increments; Offset Range: ±5°C
BNC, 2.5 mm Phono Plug, Reference Pin Tip, Ground
AC/DC 9 V Adapter (110/220 VAC, 50/60 Hz)
±0.3°C
Recorder (±2000 mV)
0.01
0.1 mV (±199.9 mV); 1 mV Beyond ±200 mV
0°C to 100°C
Automatic or Manual (0°C to 100°C)
100 Data Sets
Education Set
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers