missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Four Cell Conductivity/ATC Probes, Four-Cell

For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters

Brand:  Fisherbrand™ 93X219049

398.05 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 11560164

  • £554.00 / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters

Immersion-type cell's unique compact size is the same as a typical pH electrode—allowing use in standard electrode holder.

  • Four-cell type with glass or epoxy bodies
  • Built-in temperature sensors and 8-pin connectors
  • Cell constants of 0.1, 1.0, or 10.0cm-1

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 cm-1
Fisher Scientific accumet benchtop models XL600, XL500, XL200, & AB200 and legacy models XL60, XL50, XL30, XL20, AR60, AR50, AR30, AR20, & AB30
Glass
Immersed in deionized water
8-pin
Conductivity/ATC Probe
1 to 200 mS/cm
4 Cell Conductivity
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers