missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AccuPrime™ SuperMix II

AccuPrime SuperMix II is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.

Brand:  Invitrogen™ 12341012

 View more versions of this product

Product Code. 10554233

  • £652.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Requires : Template and primers
This item is not returnable. View return policy

Description

Description

AccuPrime SuperMix II is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half. For accurate PCR performance, AccuPrime SuperMix II sets the standard. You'll amplify your DNA targets cycle after cycle, eliminating troubleshooting and rework.

In addition to a powerful DNA polymerase, AccuPrime SuperMix I includes Platinumanti-DNA polymerase antibodies that inhibit polymerase activity, providing an automatic hot-start, and a thermal stable accessory protein that enhances specific primer-template hybridization during every cycle of PCR. This unique combination allows for success, even under previously suboptimal conditions.

AccuPrime SuperMix II is designed for amplification of genomic DNA amplicons, plasmid DNA, or cDNA templates up to 4 kb in length. Alternatively, AccuPrime SuperMix I is optimized to amplify a tighter range of targets less than 200 bp in length.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

AccuPrime SuperMix II is supplied at 2X concentration to allow approximately 50% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 200 amplification reactions of 25 μL each are provided. AccuPrime SuperMix II Components: 40 mM Tris-HCl (pH 8.4); 100 mM KCl; 3 mM MgCl2, 400 μM each dGTP, dATP, dTTP, dCTP; AccuPrime Taq DNA Polymerase; thermostable AccuPrime protein; stabilizers.

200 Reactions: 2 x 1.25 mL.
Store at -20°C in a non-frost-free freezer.
Low
2X
200 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.