missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, glacial, 99.9985% (metals basis), Thermo Scientific Chemicals

£49.95 - £598.98

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311598
View Documents
Thermo Scientific Alfa Aesar
25 mL
£49.95
25mL
Please to purchase this item. Need a web account? Register with us today!
11321598
View Documents
Thermo Scientific Alfa Aesar
010994.AE
100 mL
£150.75
100mL
Please to purchase this item. Need a web account? Register with us today!
11335819
View Documents
Thermo Scientific Alfa Aesar
500 mL
£598.98
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acetic acid is used as a solvent for purification and production of many chemical compounds eg., vinyl acetate monomer. It is a significant chemical reagent and is mainly used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as a descaling agent in households. As a food additive, it finds application as a condiment, in the pickling of vegetables and other foods and is also an acidity regulator. In analytical chemistry, for the estimation of weakly alkaline substances such as organic amides, glacial acetic acid is used. Acetic acid is used as antimicrobial preservative or acidulates in a variety of food products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

2.5
16.6°C
C2H4O2
UN2789
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
1.049 g/mL
Pungent
1.3721
Acetic acid, Glacial
64-19-7
118.1°C
MFCD00036152
14,55
Miscible with water,ethanol,acetone,toluene and immiscible with carbon disulphide.
CC(O)=O
60.05
CHEBI:15366
99.9985%
40°C (104°F)
Liquid
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL

missing translation for 'einecsNumber' : 200-580-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Acetic acid, glacial

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Acetic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only