missing translation for 'onlineSavingsMsg'
Learn More
Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™ Product Code.: 15684510

Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™

Product Code. 15684510
6 x 1 L
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
This item is not returnable. View return policy
Repeated exposure may cause skin dryness or cracking.

Product Code. 15684510

Brand: Honeywell-Chromasolv 348506X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Repeated exposure may cause skin dryness or cracking.

Acetone's luminesence intensity is dependent upon the solution components.1 The absorption of UV light by acetone results in its photolysis and the production of radials.1

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetoneShow MoreShow Less
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Density 0.791g/mL at 25°C
Boiling Point 56°C (760 mmHg)
Quantity 6 x 1 L
Refractive Index n20/D 1.359
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
Beilstein 635680
Formula Weight 58.08g/mol
Percent Purity ≥99.8%
Grade CHROMASOLV™
Chemical Name or Material Acetone, for HPLC
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.