missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™
0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, formisoton, formylic acid, collo-bueglatt, formira, aminic acid, bilorin, acide formique, myrmicyl, hydrogen carboxylic acid, methanoic acid, 2.5L
£169.00 - £526.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10468704
|
Fisher Chemical
LS120-500 |
500mL | Glass Bottle |
£169.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10678935
|
Fisher Chemical
LS120-1 |
1 L | Wide Neck Amber Bottle |
£215.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10118464
|
Fisher Chemical
LS120-212 |
2.5 L | Glass Bottle |
£526.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 1L
- Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
- Pre-mixed for ease-of-use
- Low metal ion content to ensure minimal metal contamination
- Formic acid (FA) is a common mobile phase additive that provides protons for LC/MS analysis
- FA is also known to improve the peak shapes of the resulting separation
Chemical Identifiers
75-05-8 |
Specifications
LC-MS | |
Liquid | |
75-05-8 |
0.1% Formic Acid in Acetonitrile (v/v) | |
Solvent Blends | |
Pass Test |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetonitrile with 0.1% Formic acid (v/v) for HPLC Gradient analysis
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P240-Ground/bond container and receiving equipment.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title