missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, AMD CHROMASOLV™, ≥99.9%, Honeywell Riedel-de Haën™

CHROMASOLV™ AMD,≥99.9%

£118.65 - £621.19

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15664690
View Documents
Honeywell-Chromasolv
34896-1L
1 L Glass Bottle
£118.65
1 L
Please to purchase this item. Need a web account? Register with us today!
15674690
View Documents
Honeywell-Chromasolv
34896-6X1L
6 x 1 L Glass Bottle
£621.19
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Solvents designed for high-performance thin-layer chromatography
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

-48°C (literature)
100.0000%
81.6°C
C2H3N
MFCD00001878
741857
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
High Purity
75-05-8
0.786 g/mL at 25 °C(lit.)
6°C
CH3CN
UN1648
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
>99.9%
Acetonitrile, AMD CHROMASOLV™, ≥99.9%
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1260 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile

missing translation for 'einecsNumber' : 200-835-2

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications