Learn More
Acetonitrile, Environmental Grade, 99.7+%, Thermo Scientific Chemicals
Suitable For Environmental, Pesticide Residue, and GC Analysis
£61.31 - £178.89
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11395297
|
Thermo Scientific Alfa Aesar
040972.K2 |
1 L |
£61.31
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11305307
|
Thermo Scientific Alfa Aesar
040972.M1 |
4 x 1 L |
£178.89
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11315307
|
Thermo Scientific Alfa Aesar
040972.K7 |
4 L |
£157.79
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acetonitrile is utilized as a polar aprotic solvent in organic synthesis and in the purification of butadiene. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05%, Halogenated residue 10ng/L (GC-ECD) 0.01%Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
°C to 48°C to°C to 45°C | |
Colorless | |
0.786 | |
5°C (41°F) | |
Pungent | |
1.344 | |
C2H3N | |
97 hPa (73mm Hg) at 20°C | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
Acetonitrile |
75-05-8 | |
Colorless liquid | |
80°C to 82°C | |
525°C (977°F) | |
Liquid | |
>99.7% | |
MFCD00001878 | |
741857 | |
Miscible with water and almost all organic solvents. | |
CC#N | |
41.053 | |
CHEBI:38472 | |
Environmental |
Safety and Handling
Keep away from sources of ignition - No Smoking. Wear suitable protective clothing and gloves.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
H225-H302+H312+H332-H319
missing translation for 'dotInformation' : DOT Class: 3, Packaging Group: II
missing translation for 'einecsNumber' : 200-835-2
missing translation for 'rtecsNumber' : AL7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.