missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, Honeywell
£140.05 - £10681.11
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow More |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15655900
|
Honeywell Research Chemicals
60004-1L |
1 L |
£140.05
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15665900
|
Honeywell Research Chemicals
60004-2.5L |
2.5 L |
£325.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15615910
|
Honeywell Research Chemicals
60004-5L |
5 L |
£447.54
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15685900
|
Honeywell Research Chemicals
60004-25L |
25 L | N/A | ||||||
15675900
|
Honeywell Research Chemicals
60004-20KG |
20 kg | N/A | ||||||
15645900
|
Honeywell Research Chemicals
60004-150KG |
150 kg | N/A | ||||||
Description
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow More | |
CHEBI:38472 | |
CC#N |
Specifications
75-05-8 | |
0.16 | |
973°F | |
CH3CN | |
72.8 mmHg (20°C) | |
741857 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05g/mol | |
Acetonitrile |
0.786g/mL at 25°C | |
2°C (35.6°F) | |
C2H3N | |
MFCD00001878 | |
UN1648 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.5% (GC) |
Safety and Handling
P210-P280-P305 + P351 + P338
Danger
missing translation for 'einecsNumber' : 200-835-2
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Acetonitrile
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : acetonitrile
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetonitrile, Honeywell