missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals Product Code.: 15492857

Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals

Product Code. 15492857
1 L
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
This item is not returnable. View return policy

Product Code. 15492857

Brand: Thermo Scientific Alfa Aesar 047138.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solvent for LC-MS analysis

Solvent for LC-MS analysisAcetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow MoreShow Less
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C to -45°C
Color Colorless
Density 0.786 g/mL
Boiling Point 80°C to 82°C
Quantity 1 L
Flash Point 5°C (41°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.344
UN Number UN1648
Beilstein 741857
Merck Index 14,70
Solubility Information Miscible with water,methanol,methyl acetate,ethyl acetate and chloroform. Immiscible with saturated hydrocarbons.
Formula Weight 41.05
Percent Purity 99.8%
Grade LC-MS
Chemical Name or Material Acetonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP14L709 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog Number047138.M1,047138.K7,047138.M6,047138.K2
Lot NumberP14L708 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog Number047138.M1,047138.K7,047138.M6,047138.K2
Lot NumberN28L713 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog Number047138.M1,047138.K7,047138.M6,047138.K2
Lot NumberM30L712 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog Number047138.M1,047138.K7,047138.M6,047138.K2
Lot NumberY15K612 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog Number047138.M1,047138.K7,047138.M6,047138.K2
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Acetonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.