missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation ACQUITY HSS PFP Column

ACQUITY HSS PFP columns are fully scalable and transferable to HPLC separations.

Brand:  Waters Corporation 186005964

Encompass

 View more versions of this product

Product Code. 17123392

  • £1042.50 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ACQUITY UPLC HSS PFP is a non-endcapped silica-based column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase.
Specifications

Specifications

2.1 mm
18000 psi (1240 Bar)
1.8 μm
1/Ea.
No
30 mm
Spherical
100 Å
L43
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers