missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation ACQUITY UPLC BEH HILIC Column
ACQUITY BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds.
Brand: Waters Corporation 186004677
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
This item is not returnable.
View return policy
Description
- These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
- More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
Specifications
3 mm | |
18000 psi (1240 Bar) | |
1.7 μm | |
1/Ea. | |
No |
150 mm | |
Spherical | |
130 Å | |
L3 | |
Column |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction