Learn More
Waters Corporation ACQUITY UPLC BEH HILIC Column
ACQUITY BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds.
£1072.50 - £3375.00
Specifications
Max. Pressure | 18000 psi (1240 Bar) |
---|---|
Particle Shape | Spherical |
Particle Size | 1.7 μm |
Pore Size | 130 Å |
USP Type | L3 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
13175129
|
Waters Corporation
186003459 |
150 mm | 1 mm | 1/Ea. |
£1215.00
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
12613456
|
Waters Corporation
186003460 |
50 mm | 2.1 mm | 1/Ea. |
£1072.50
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
10260205
|
Waters Corporation
186003461 |
100 mm | 2.1 mm | 1/Ea. |
£1132.50
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13195129
|
Waters Corporation
186003462 |
150 mm | 2.1 mm | 1/Ea. |
£1215.00
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13155129
|
Waters Corporation
186003457 |
50 mm | 1 mm | 1/Ea. |
£1072.50
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13165129
|
Waters Corporation
186003458 |
100 mm | 1 mm | 1/Ea. |
£1132.50
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
10493865
|
Waters Corporation
176001089 |
50 mm | 1 mm | 3/Pk. |
£3000.00
Pack of 3 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13129919
|
Waters Corporation
176001090 |
100 mm | 1 mm | 3/Pk. |
£3180.00
Pack of 3 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13139919
|
Waters Corporation
176001091 |
150 mm | 1 mm | 3/Pk. |
£3375.00
Pack of 3 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13149919
|
Waters Corporation
176001092 |
50 mm | 2.1 mm | 3/Pk. |
£3000.00
Pack of 3 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13159919
|
Waters Corporation
176001093 |
100 mm | 2.1 mm | 3/Pk. |
£3180.00
Pack of 3 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13169919
|
Waters Corporation
176001094 |
150 mm | 2.1 mm | 3/Pk. |
£3375.00
Pack of 3 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13200020
|
Waters Corporation
176001811 |
100 mm | 3 mm | 3/Pk. |
£3180.00
Pack of 3 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13263820
|
Waters Corporation
186004676 |
100 mm | 3 mm | 1/Ea. |
£1132.50
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13290010
|
Waters Corporation
176001810 |
50 mm | 3 mm | 3/Pk. |
£3000.00
Pack of 3 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13210020
|
Waters Corporation
176001812 |
150 mm | 3 mm | 3/Pk. |
£3375.00
Pack of 3 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13253820
|
Waters Corporation
186004675 |
50 mm | 3 mm | 1/Ea. |
£1072.50
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
13273820
|
Waters Corporation
186004677 |
150 mm | 3 mm | 1/Ea. |
£1215.00
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
17113212
|
Waters Corporation
186005608 |
75 mm | 2.1 mm | 1/Ea. |
£1110.00
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
17163212
|
Waters Corporation
186005613 |
75 mm | 3 mm | 1/Ea. |
£1110.00
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
Description
- These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
- More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
Specifications
18000 psi (1240 Bar) | |
1.7 μm | |
L3 | |
Column |
Spherical | |
130 Å | |
No |