missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acro™ 50 Vent Devices with PTFE Membrane

Disposable filters ideal for the sterile venting of small containers and as an in-line gas filter.

Brand:  Cytiva (formerly Pall Lab) 4250

Additional Details : Weight : 3.26000kg

 View more versions of this product

Product Code. 17114291

  • £710.00 / Pack of 72
Estimated Shipment: 07-10-2024
to see stock.
Add to basket

Description

Description

  • Contain a hydrophobic PTFE membrane
  • Large effective filter area provides high flow rates and filters can be autoclaved several times ensuring low costs
Specifications

Specifications

Autoclavable
0.2 μm
19.6 cm2
Stepped Hose Barb Connection
50 mm
PTFE
Non-sterile
Polypropylene
72/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers