missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acro™ 50 Vent Devices with PTFE Membrane

Disposable filters ideal for the sterile venting of small containers and as an in-line gas filter.

Brand:  Cytiva (formerly Pall Lab) 4400

212.77 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.



Additional Details : Weight : 0.89300kg

 View more versions of this product

Product Code. 17134301

  • £519.00 / Pack of 18
In Stock
to see stock.
Add to basket

Description

Description

  • Contain a hydrophobic PTFE membrane
  • Large effective filter area provides high flow rates and filters can be autoclaved several times ensuring low costs
Specifications

Specifications

Autoclavable
0.2 μm
19.6 cm2
1/8 in. MNPT Connection
50 mm
PTFE
Non-sterile
Polypropylene
18/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers