missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Acro™ 50 Vent Devices with PTFE Membrane

Disposable filters ideal for the sterile venting of small containers and as an in-line gas filter.

Brand:  Cytiva 4401

268.85 GBP valid until 2025-03-14
Use promo code "24142" to get your promotional price.


 View more versions of this product

Product Code. 17144301

  • £283.00 / Pack of 18
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Contain a hydrophobic PTFE membrane
  • Large effective filter area provides high flow rates and filters can be autoclaved several times ensuring low costs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
0.2 μm
19.6 cm2
3/8 in. Straight Pipe Connection
50 mm
PTFE
Non-sterile
Polypropylene
18/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Acro™ 50 Vent Devices with PTFE Membrane > 0.2 μm, 3/8 in. straight pipe; 18/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.