missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acro™ 50 Vent Devices with PTFE Membrane

Disposable filters ideal for the sterile venting of small containers and as an in-line gas filter.

Brand:  Cytiva (formerly Pall Lab) 4258

Additional Details : Weight : 0.87400kg

 View more versions of this product

Product Code. 17154291

  • £288.00 / Pack of 18
Estimated Shipment: 30-08-2024
to see stock.
Add to basket

Description

Description

  • Contain a hydrophobic PTFE membrane
  • Large effective filter area provides high flow rates and filters can be autoclaved several times ensuring low costs
Specifications

Specifications

Autoclavable
1 μm
19.6 cm2
Stepped Hose Barb Connection
50 mm
PTFE
Non-sterile
Polypropylene
18/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers