Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PF Syringe Filters with Supor™ Membrane, Sterile
These syringe filters are ideal for biological and general research.
Brand:  Cytiva (formerly Pall Lab) 4658
Additional Details : Weight : 0.47500kg
 View more versions of this product
Product Code. 17174381
  • £159.96 / Pack of 50
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
  • Syringe filters perform sample clean-up and preparation, ensuring the purest samples are analyzed in liquid chromatography, mass spectrometry, and ion chromatography applications
  • Superior flow rates and higher throughputs than competitive devices
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
PES
Sterile
32 mm
0.8/0.2 μm
50/Pk.
Product Suggestions
Product Suggestions
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote