missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filter with Polypropylene Housing

Syringe Filter for use in cell culture and other sensitive biological research.

Brand:  Cytiva (formerly Pall Lab) AP-4529

2499.93 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.


 View more versions of this product

Product Code. 17164681

  • £4065.00 / Pack of 1000
Estimated Shipment: 15-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • This product is not approved for use in medical, clinical, surgical or other patient applications
  • Meets aqueous sample filtration needs
  • Available with GxF multi-layered glass fiber prefilter, providing two to four times the throughput of standard glass fiber prefilter devices, and allowing quick and easy filtration of your most difficult-to-filter samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glass Fiber, Borosilicate Glass Fiber
Non-sterile
<125 μL
1000/Pk.
25 mm
1 μm
3.9 cm2
Polypropylene
55°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers