missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with PVDF Membrane

Hydrophilic membrane compatible with a wide variety of solvents.

Brand:  Cytiva (formerly Pall Lab)

Additional Details : Weight : 3.56250kg

 View more versions of this product

Product Code. 17124331

  • £2760.00 / Pack of 1000
Estimated Shipment: 05-09-2024
to see stock.
Add to basket

Description

Description

  • Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
Specifications

Specifications

PVDF
Non-sterile
<100 μL
1000/Pk.
25 mm
0.45 μm
3.9 cm2
Polypropylene
82°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers