missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with PVDF Membrane
Hydrophilic membrane compatible with a wide variety of solvents.
Brand: Cytiva (formerly Pall Lab) 4408
Additional Details : Weight : 0.85500kg
Description
- Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
Specifications
PVDF | |
Non-sterile | |
<100 μL | |
200/Cs. | |
82°C |
0.45 μm | |
3.9 cm2 | |
Polypropylene | |
50/Pk. | |
25 mm |