missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with wwPTFE Membrane

Universal membrane syringe filters for all of your analytical filtration requirements.

Brand:  Cytiva (formerly Pall Lab) 4914

Additional Details : Weight : 0.43800kg

 View more versions of this product

Product Code. 17134451

  • £338.00 / Pack of 50
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

wwPTFE
Non-sterile
<100 μL
200/Cs.
55°C
0.45 μm
2.8 cm2
Polypropylene
50/Pk.
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers