missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with wwPTFE Membrane
Universal membrane syringe filters for all of your analytical filtration requirements.
Brand: Cytiva (formerly Pall Lab) 4929
Additional Details : Weight : 3.21100kg
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
wwPTFE | |
Non-sterile | |
<100 μL | |
1000/Pk. | |
25 mm |
0.2 μm | |
2.8 cm2 | |
Polypropylene | |
55°C |