missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with wwPTFE Membrane and Minispike Outlet

Universal membrane syringe filters for all of your analytical filtration requirements.

Brand:  Cytiva (formerly Pall Lab) 2403

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 17134261

  • £1570.00 / Pack of 1000
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

wwPTFE
Non-sterile
<14 μL
1000/Pk.
55°C
0.45 μm
1 cm2
Polypropylene
Minispike Outlet
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers