Learn More
Cytiva (formerly Pall Lab) AcroPak™ 1000 Capsules with Supor™ Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
Brand:  Cytiva (formerly Pall Lab) 12999
Additional Details : Weight : 0.19000kg
 View more versions of this product
Product Code. 17174251
  • £216.00 / Each
Estimated Shipment: 30-08-2024
to see stock.
Description
Description
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
  • Designed for research applications only
  • Gamma-irradiated
Specifications
Specifications
Hydrophilic PES
Sterile
Stepped Hose Barb
0.1/0.1 μm
1000 cm2
1/Ea.