missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 1000 Capsules with Supor™ Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
£178.00 - £216.00
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Sterility | Sterile |
Filtration Area | 1000 cm2 |
Outlet Style | Stepped Hose Barb |
Quantity | 1/Ea. |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
17114251
|
Cytiva (formerly Pall Lab)
12992 |
0.8/0.2 μm |
£183.70
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17134251
|
Cytiva (formerly Pall Lab)
12994 |
0.8/0.45 μm |
£178.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17154251
|
Cytiva (formerly Pall Lab)
12996 |
0.2/0.2 μm |
£186.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17174251
|
Cytiva (formerly Pall Lab)
12999 |
0.1/0.1 μm |
£216.00
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
- Designed for research applications only
- Gamma-irradiated
Specifications
Hydrophilic PES | |
1000 cm2 | |
1/Ea. |
Sterile | |
Stepped Hose Barb |