Learn More
Cytiva (formerly Pall Lab) AcroPak™ 1500 Capsules with Supor™ EKV Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
Brand:  Cytiva (formerly Pall Lab) 12466
Additional Details : Weight : 0.26800kg
 View more versions of this product
Product Code. 17124241
  • £214.00 / Each
Estimated Shipment: 17-10-2024
to see stock.
Description
Description
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
  • Designed for research applications only
  • Gamma-irradiated
Specifications
Specifications
Hydrophilic PES
Sterile
1 to 1.5 in. Sanitary Flange
0.2 μm
1500 cm2
1/Ea.