missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 1500 Capsules with Supor™ EKV Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
£214.00
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Pore Size | 0.2 μm |
Sterility | Sterile |
Filtration Area | 1500 cm2 |
Quantity | 1/Ea. |
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
17134241
|
Cytiva (formerly Pall Lab)
12467 |
1/2 in. Hose Barb |
£214.00
Each |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17124241
|
Cytiva (formerly Pall Lab)
12466 |
1 to 1.5 in. Sanitary Flange |
£214.00
Each |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
- Designed for research applications only
- Gamma-irradiated
Specifications
Hydrophilic PES | |
Sterile | |
1/Ea. |
0.2 μm | |
1500 cm2 |