Learn More
Cytiva AcroPak™ 1500 Capsules with Supor™ Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
£211.00
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Sterility | Sterile |
Filtration Area | 1500 cm2 |
Outlet Style | 1/2 in. Hose Barb |
Quantity | 1/Ea. |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
17174241
|
Cytiva
12675 |
0.8/0.2 μm |
£211.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17184241
|
Cytiva
12686 |
0.2/0.2 μm |
£211.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
- Designed for research applications only
- Gamma-irradiated
Specifications
Hydrophilic PES | |
1500 cm2 | |
1/Ea. |
Sterile | |
1/2 in. Hose Barb |