missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 200 Capsules with Supor™ EKV Membrane
AcroPak capsules designed for research applications only.
Brand: Cytiva (formerly Pall Lab) 12094
This item is not returnable.
View return policy
Description
- Efficient processing of liquid volumes up to 20 L
- Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
Specifications
Hydrophilic PES | |
Sterile | |
Stepped Hose Barb Inlet, Stepped Hose Barb Outlet with Filling Bell |
0.2 μm | |
200 cm2 | |
3/Pk. |