missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 200 Capsules with Supor™ EKV Membrane
AcroPak capsules designed for research applications only.
£250.00
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
17144221
|
Cytiva (formerly Pall Lab)
12094 |
Stepped Hose Barb Inlet, Stepped Hose Barb Outlet with Filling Bell |
£250.00
Pack of 3 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17154221
|
Cytiva (formerly Pall Lab)
12095 |
1/2 in. Sanitary Flange |
£250.00
Pack of 3 |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Efficient processing of liquid volumes up to 20 L
- Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
Specifications
Hydrophilic PES | |
0.2 μm | |
3/Pk. |