missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 200 Capsules with Supor™ EKV Membrane
AcroPak capsules designed for research applications only.
Brand: Cytiva (formerly Pall Lab) 12095
Additional Details : Weight : 0.38850kg
Description
- Efficient processing of liquid volumes up to 20 L
- Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
Specifications
Hydrophilic PES | |
Non-sterile | |
1/2 in. Sanitary Flange |
0.2 μm | |
220 cm | |
3/Pk. |