missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 200 Capsules with Supor™ Membrane
AcroPak capsules designed for research applications only.
£232.00 - £316.00
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Pore Size | 0.8/0.2 μm |
Sterility | Sterile |
Filtration Area | 200 cm2 |
Quantity | 3/Pk. |
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
17194241
|
Cytiva (formerly Pall Lab)
12941 |
Stepped Hose Barb |
£316.00
Pack of 3 |
Estimated Shipment: 14-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17134221
|
Cytiva (formerly Pall Lab)
12093 |
1/4 in. MNPT Inlet, Stepped Hose Barb Outlet |
£232.00
Pack of 3 |
Estimated Shipment: 14-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Efficient processing of liquid volumes up to 20 L
- Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Hydrophilic PES | |
Sterile | |
3/Pk. |
0.8/0.2 μm | |
200 cm2 |