Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for DNA Purification
Optimized for a variety of applications requiring larger per well volume processing.
Brand:  Cytiva (formerly Pall Lab) 8032
Additional Details : Weight : 0.19200kg
 View more versions of this product
Product Code. 17164601
  • £295.00 / Pack of 10
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Borosilicate Glass w/o Binder
Polypropylene Housing
350 μL
Non-sterile
10/Pk.