Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for DNA Purification
Optimized for a variety of applications requiring larger per well volume processing.
£155.00 - £295.00
Specifications
Material (Membrane) | Borosilicate Glass w/o Binder |
---|---|
Sterility | Non-sterile |
Housing Material | Polypropylene Housing |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
17164601
|
Cytiva (formerly Pall Lab)
8032 |
350 μL |
£295.00
Pack of 10 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||||
17144621
|
Cytiva (formerly Pall Lab)
8132 |
1 mL |
£155.00
Pack of 5 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Borosilicate Glass w/o Binder | |
Non-sterile | |
Polypropylene Housing |