Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Optimized for a variety of applications requiring larger per well volume processing.
Brand:  Cytiva (formerly Pall Lab) 8584
Additional Details : Weight : 0.45400kg
 View more versions of this product
Product Code. 17114641
  • £202.00 / Pack of 10
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
wwPTFE
Non-sterile
10/Pk.
0.45 μm
Solvent Filtration
350 μL