Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Optimized for a variety of applications requiring larger per well volume processing.
£152.00 - £327.00
Product Code | Brand | Volume (Metric) | Pore Size | Material (Membrane) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Pore Size | Material (Membrane) | Price | Quantity & Availability | |||
17134611
|
Cytiva (formerly Pall Lab)
8047 |
350 μL | 0.2 μm | PTFE |
£301.00
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
17114621
|
Cytiva (formerly Pall Lab)
8129 |
1 mL | 0.45 μm | Supor (PES) |
£161.00
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
17124621
|
Cytiva (formerly Pall Lab)
8130 |
1 mL | 1.2 μm | Supor (PES) |
£201.00
Pack of 5 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||
17144611
|
Cytiva (formerly Pall Lab)
8048 |
350 μL | 0.45 μm | PTFE |
£291.00
Pack of 10 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||
17174631
|
Cytiva (formerly Pall Lab)
8247 |
2 mL | 0.2 μm | PTFE |
£227.00
Pack of 5 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||
17164621
|
Cytiva (formerly Pall Lab)
8147 |
1 mL | 0.2 μm | PTFE |
£306.00
Pack of 5 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||
17174621
|
Cytiva (formerly Pall Lab)
8148 |
1 mL | 0.45 μm | PTFE |
£327.00
Pack of 5 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||
17104641
|
Cytiva (formerly Pall Lab)
8582 |
350 μL | 0.2 μm | wwPTFE |
£202.00
Pack of 10 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||
17144641
|
Cytiva (formerly Pall Lab)
8682 |
1 mL | 0.2 μm | wwPTFE |
£152.00
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
17154641
|
Cytiva (formerly Pall Lab)
8684 |
1 mL | 0.45 μm | wwPTFE |
£163.00
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
17114641
|
Cytiva (formerly Pall Lab)
8584 |
350 μL | 0.45 μm | wwPTFE |
£202.00
Pack of 10 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
17184631
|
Cytiva (formerly Pall Lab)
8248 |
2 mL | 0.45 μm | PTFE |
£177.00
Pack of 5 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||
17184641
|
Cytiva (formerly Pall Lab)
8782 |
2 mL | 0.65/0.2 μm | wwPTFE |
£182.00
Pack of 5 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||
17194641
|
Cytiva (formerly Pall Lab)
8784 |
2 mL | 0.45 μm | wwPTFE |
£304.44
Pack of 5 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||
Beschreibung
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Spezifikation
Non-sterile | |
Solvent Filtration |