missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration

Optimized for a variety of applications requiring larger per well volume processing.

Brand:  Cytiva (formerly Pall Lab) 8247

Additional Details : Weight : 1.11150kg

 View more versions of this product

Product Code. 17174631

  • £227.00 / Pack of 5
Estimated Shipment: 11-09-2024
to see stock.
Add to basket

Description

Description

  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

PTFE
Non-sterile
5/Pk.
0.2 μm
Solvent Filtration
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers