Learn More
Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents
£111.00 - £1310.00
Chemical Identifiers
CAS | 79-06-1 |
---|---|
Molecular Formula | C3H5NO |
Molecular Weight (g/mol) | 71.08 |
MDL Number | MFCD00008032 |
InChI Key | HRPVXLWXLXDGHG-UHFFFAOYSA-N |
PubChem CID | 6579 |
ChEBI | CHEBI:28619 |
IUPAC Name | prop-2-enamide |
SMILES | NC(=O)C=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10562595
|
Fisher BioReagents
BP170-100 |
100 g | Poly Bottle |
£111.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10235203
|
Fisher BioReagents
BP170-500 |
500 g | Poly Bottle |
£176.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10502605
|
Fisher BioReagents
BP170-5 |
5 kg | Poly Pail |
£1310.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Acrylamide is 3X recrystallized to obtain a high-purity crystalline material suitable for electrophoretic separation of proteins and nucleic acids.Chemical Identifiers
79-06-1 | |
71.08 | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
CHEBI:28619 | |
NC(=O)C=C |
C3H5NO | |
MFCD00008032 | |
6579 | |
prop-2-enamide |
Specifications
Solid | |
79-06-1 | |
82°C | |
White | |
DNase free | |
C3H5NO | |
MFCD00008032 | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enamide | |
6579 | |
71.08 | |
DNase-, RNase- and Protease-Free |
0.002% max. | |
125°C | |
2.50μmhos max. | |
6.5 to 8.0 | |
≥99.9 % | |
CH2:CHCONH2 | |
15, 120 | |
NC(=O)C=C | |
71.08 | |
CHEBI:28619 | |
≥99.9% | |
Acrylamide |
Safety and Handling
WARNING!
Emergency Overview
Toxic if swallowed. May cause cancer. Harmful if absorbed through skin or if inhaled. May impair fertility. May cause heritable genetic damage. May cause an allergic skin reaction. Irritating to eyes and skin. Toxic: danger of serious damage to health by prolonged exposure through inhalation, in contact with. May cause central nervous system effects. Do not breathe dust. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Do not get in eyes, on skin, or on clothing. Keep away from heat, sparks and open flame - No smoking.
NFPA
Health:2
Flammability:2
Instability:2
missing translation for 'dotInformation' : DOT Class 6.1, : Poison
Recommended Storage : RT
- Acrylamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H372-Causes damage to organs.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361f-Suspected of damaging fertility.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.