Learn More
Acrylonitrile, 99+%, Thermo Scientific Chemicals
£18.60 - £219.00
Chemical Identifiers
CAS | 107-13-1 |
---|---|
Molecular Formula | C3H3N |
Molecular Weight (g/mol) | 53.06 |
MDL Number | MFCD00001927 |
InChI Key | NLHHRLWOUZZQLW-UHFFFAOYSA-N |
Synonym | acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox |
PubChem CID | 7855 |
ChEBI | CHEBI:28217 |
IUPAC Name | prop-2-enenitrile |
SMILES | C=CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10386500
|
Thermo Scientific Acros
|
100 mL | Glass bottle |
£18.60
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10163720
|
Thermo Scientific Acros
|
1 L | Glass bottle |
£36.60
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686922
|
Thermo Scientific Acros
|
2.5 L | Glass bottle |
£67.80
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10145833
|
Thermo Scientific Acros
|
10 L | Plastic drum |
£219.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-13-1 | |
53.06 | |
NLHHRLWOUZZQLW-UHFFFAOYSA-N | |
7855 | |
prop-2-enenitrile |
C3H3N | |
MFCD00001927 | |
acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox | |
CHEBI:28217 | |
C=CC#N |
Specifications
107-13-1 | |
100.0 | |
0.8000g/mL | |
77.3°C | |
Authentic | |
C3H3N | |
1.3900 to 1.3920 | |
02, 400 | |
15, 122 | |
acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox | |
NLHHRLWOUZZQLW-UHFFFAOYSA-N | |
prop-2-enenitrile | |
53.06 | |
CHEBI:28217 | |
99+% | |
Acrylonitrile |
99.0 | |
-83.5°C | |
5.9 to 7.6 (5% aq. soln.) | |
−0.2°C | |
99% min. (GC) | |
H2C=CHCN | |
MFCD00001927 | |
01,14; 07,4; 13,317 | |
0.8 | |
Solubility in water: 73.5g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone | |
C=CC#N | |
35 to 45ppm MEHQ | |
7855 | |
53.06 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause cancer.
May cause respiratory irritation.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Toxic
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-466-5
- Acrylonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H361-Suspected of damaging fertility or the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only