missing translation for 'onlineSavingsMsg'
Learn More
Acrylonitrile, 99+%, Thermo Scientific Chemicals Product Code.: 10163720

Acrylonitrile, 99+%, Thermo Scientific Chemicals

Product Code. 10163720
1 L, Glass bottle
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10163720

Brand: Thermo Scientific Acros 149630010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-13-1
Molecular Formula C3H3N
Molecular Weight (g/mol) 53.06
MDL Number MFCD00001927
InChI Key NLHHRLWOUZZQLW-UHFFFAOYSA-N
Synonym acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox
PubChem CID 7855
ChEBI CHEBI:28217
IUPAC Name prop-2-enenitrile
SMILES C=CC#N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -83.5°C
Density 0.8000g/mL
pH 5.9 to 7.6 (5% aq. soln.)
Boiling Point 77.3°C
Flash Point −0.2°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCN
Refractive Index 1.3900 to 1.3920
Quantity 1 L
Beilstein 02, 400
Fieser 01,14; 07,4; 13,317
Merck Index 15, 122
Specific Gravity 0.8
Solubility Information Solubility in water: 73.5g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Stabilizer 35 to 45ppm MEHQ
Formula Weight 53.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Acrylonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463137 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number149630100,149630025,149630010,149631000
Lot NumberA0449386 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog Number149630100,149630025,149630010,149631000
Lot NumberA0447177 Certificate TypeCertificate of Analysis Date02/11/2023 Catalog Number149630100,149630025,149630010,149631000
Lot NumberA0456598 Certificate TypeCertificate of Analysis Date27/09/2023 Catalog Number149630100,149630025,149630010,149631000
Lot NumberA0397765 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number149630100,149630025,149630010,149631000
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acrylonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acrylonitrile, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.