missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapter for 250 mL Rectangular Bucket in Rotor FA - 6 x 250

Adapts a wide range of centrifuge tubes for use in Rotor FA-6x250.

Brand:  Eppendorf™ 5920765000

 View more versions of this product

Product Code. 16262411

  • £301.00 / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Adapts a wide range of centrifuge tubes to fit in Rotor FA - 6 x 250
  • Set of 2 pieces
TRUSTED_SUSTAINABILITY
Specifications

Specifications

9 x FACS™ Round Tubes, 5 mL
For 250 mL rectangular bucket in Rotor FA-6x250
9
Set of 2 pieces
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers