Learn More
Eppendorf™ Adapter for 250 mL Rectangular Bucket in Rotor FA - 6 x 250
Adapts a wide range of centrifuge tubes for use in Rotor FA-6x250.
£286.00 - £301.00
Specifications
Description | For 250 mL rectangular bucket in Rotor FA-6x250 |
---|---|
No. per Pack | Set of 2 pieces |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
16202421
|
Eppendorf™
5920769006 |
7 x 10 mL Oak Ridge Round Centrifuge Tubes |
£286.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16212411
|
Eppendorf™
|
1 x 50 mL Conical Tube |
£286.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16212421
|
Eppendorf™
|
7 x 16 mL Oak Ridge Round Centrifuge Tubes |
£301.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16222411
|
Eppendorf™
|
4 x Eppendorf Tubes™ 5.0 mL and 15 mL conical tubes. (Universal adapters required for 5 mL tubes.) |
£286.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16222421
|
Eppendorf™
5920771000 |
1 x 50 mL Oak Ridge Round Centrifuge Tube |
£286.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16232411
|
Eppendorf™
5920762001 |
7 x Blood Collection Round Tubes: D16, 5.5 to 12.0 mL |
£286.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16242411
|
Eppendorf™
5920763008 |
8 x Blood Collection Round Tubes: D13, 2.6 to 8.0 mL |
£301.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16262411
|
Eppendorf™
5920765000 |
9 x FACS™ Round Tubes, 5 mL |
£301.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16272411
|
Eppendorf™
5920766007 |
1 x 50 mL Skirted Tubes |
£286.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16282411
|
Eppendorf™
5920767003 |
3 x 30 mL Oak Ridge Round Centrifuge Tubes |
£301.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16292411
|
Eppendorf™
5920768000 |
1 x 85 mL Oak Ridge Round Centrifuge Tube |
£286.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Adapts a wide range of centrifuge tubes to fit in Rotor FA - 6 x 250
- Set of 2 pieces
Specifications
For 250 mL rectangular bucket in Rotor FA-6x250 | |
Set of 2 pieces |