Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
Brand: Eppendorf™ 5920700006
271.70 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for S-4xUniversal rotor buckets
- Accommodates selected array of tubes, bottles, or plates
Specifications
Flat Bottom | |
98 mm | |
S-4xUniversal Rotor | |
2 |
1 x 1 L Nalgene Bottle | |
185 mm | |
1 |