Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
£286.00 - £432.72
Specifications
For Use With (Equipment) S-4xUniversal Rotor
No. per Pack 2
View More Specs
Products 12
Product Code Brand Capacity Tube Type No. of Places Price Quantity & Availability  
Product Code Brand Capacity Tube Type No. of Places Price Quantity & Availability  
15810937
View Documents
Eppendorf™
5910764006
7 x 50 mL Skirted Tubes, 7 x 30 mL Oak Ridge Tubes Skirted Bottom, Oak Ridge 7/7
£301.00
Pack of 2
15820937
View Documents
Eppendorf™
5910769008
9 x 50 mL Conical Tubes, 1 Microplate Conical 9/1
£344.00
Pack of 2
15758112
View Documents
Eppendorf™
5920700006
1 x 1 L Nalgene Bottle Flat Bottom 1
£286.00
Pack of 2
In Stock
to see stock.
Add to basket
15890927
View Documents
Eppendorf™
5910760000
1 x 500 mL Conical Bottle Conical 1
£293.17
Pack of 2
15870927
View Documents
Eppendorf™
5910757000
1 x 750 mL Flat-bottom Bottle Flat Bottom 1
£301.00
Pack of 2
15840927
View Documents
Eppendorf™
5910753004
5 x MTP/DWP/PCR Plates PCR 5
£293.17
Pack of 2
15820927
View Documents
Eppendorf™
5910751001
7 x 50 mL Conical Tubes, 1 x 96/384-well Microplate, 1 x 250 mL Flat-bottom Bottle, 1 x 175 to 225 mL Conical Bottle Flat/Conical Bottom 7/1/1/1
£432.72
Pack of 2
15880927
View Documents
Eppendorf™
5910758006
46 tubes of 12 x 75mm or 5ml, Round Bottom Tubes (FACS) Round Bottom (FACS) 46
£307.22
Pack of 2
15830927
View Documents
Eppendorf™
5910752008
16 x 5 mL Microtubes, 16 x 15 mL Conical Tubes, 1 MTP/DWP Microtube, Conical 16/16/1
£293.17
Pack of 2
15800937
View Documents
Eppendorf™
5910762003
21 x 9 mL Round Bottom Tubes Round Bottom 21
£301.00
Pack of 2
15850927
View Documents
Eppendorf™
5910754000
26 x 7.5 to 12 mL Round Bottom Tubes Round Bottom 26
£301.00
Pack of 2
15860927
View Documents
Eppendorf™
5910755007
30 x 4 to 8 mL Round Bottom Tubes Round Bottom 30
£293.17
Pack of 2
Description
Description
  • Suitable for S-4xUniversal rotor buckets
  • Accommodates selected array of tubes, bottles, or plates
Specifications
Specifications
S-4xUniversal Rotor
2