Learn More
Thermo Scientific™ Adapters for HIGHConic III Rotor
Assorted adapters to be used with the HIGHConic III rotor
£116.97 - £252.59
Product Code | Brand | Capacity | Max. Tube Diameter | Max. Tube Length | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Max. Tube Diameter | Max. Tube Length | Price | Quantity & Availability | |||
15798139
|
Thermo Scientific™
75005802 |
38 mL | 25.5 mm | 110 mm |
£144.09
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15708149
|
Thermo Scientific™
75005803 |
16 mL | 18 mm | 123 mm |
£139.86
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15738578
|
Thermo Scientific™
75005804 |
12 mL | 16 mm | 96 mm |
£144.09
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15718149
|
Thermo Scientific™
75005805 |
6.5 mL | 13.5 mm | 114 mm |
£144.09
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15728149
|
Thermo Scientific™
75005806 |
3.5 mL | 11 mm | 100 mm |
£149.85
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15753569
|
Thermo Scientific™
75005807 |
1.5/2.0 mL | 11 mm | 40 mm |
£144.09
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15105273
|
Thermo Scientific™
75005808 |
8 x 15 mL | 12 mm | 17 mm |
£252.59
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15271026
|
Thermo Scientific™
75005770 |
5 mL | 17 mm | 100 mm |
£116.97
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Accommodate conical tubes in the 50mL buckets for use with the Thermo Scientific HIGHConic III fixed angle rotor.
- Adapters to run various tubes in HIGHConic III rotor
Specifications
Rotor Adapter |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Adapters for HIGHConic III Rotor