Learn More
Thermo Scientific™ Adapters for Thermo Scientific™ MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
PCR Tube and Microtube Adapters
Brand:  Thermo Scientific™ 75005754
 View more versions of this product
Product Code. 15173263
  • £262.76 / Each
Explore more special offers
Description
Description
Adapters for Thermo Scientific MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Adapter
0.25 mL
0.25 mL
Microtube
MicroClick 30 x 2 and MicroClick 24 x 2 rotor
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Adapters for Thermo Scientific™ MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor > 0.25mL Microtube