Learn More
Thermo Scientific™ Additional Activated Carbon Filter
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Brand:  Thermo Scientific™ 50073773
1602.84 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 04
Additional Details : Weight : 12.32100kg
 View more versions of this product
Product Code. 10003084
  • £1755.39 / Each
In Stock
to see stock.
Add to basket
Description
Description
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
  • Additional Activated Carbon Filter
  • Fittings Included.
  • Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Specifications
Specifications
EN 12469
Activated Carbon
Retrofit to 1.2/1.5 m Cabinets
Filter
Fittings