missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Adenosine 5'-diphosphate, disodium salt hydrate, 98% Product Code.: 10143940

Thermo Scientific Chemicals Adenosine 5'-diphosphate, disodium salt hydrate, 98%

Product Code. 10143940
1 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10143940

Brand: Thermo Scientific Chemicals 164670010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrateShow MoreShow Less
PubChem CID 85315
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Absorbance 0.76 to 0.80 (A250/A260), 0.15 to 0.17 (A280/A260)
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information (5% w/v in water) Clear colorless
Spectrophotometry 97.5 to 102.5% (ADP-Na2H,on dry basis) (calculated) 82% min. (ADP,on as is basis) (lambda max. 260nm) molar absorptivity 15000+/-2%,pH 7
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
Formula Weight 471.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenosine 5′-diphosphate, disodium salt hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0448796 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number164672500,164670050,164670010
Lot NumberA0448796 Certificate TypeCertificate of Analysis Date02/01/2024 Catalog Number164672500,164670050,164670010
Lot NumberA0406161 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number164672500,164670050,164670010
Lot NumberA0417736 Certificate TypeCertificate of Analysis Date06/04/2022 Catalog Number164672500,164670050,164670010
Lot NumberA0439236 Certificate TypeCertificate of Analysis Date28/02/2022 Catalog Number
Product Identifier
  • Adenosine-5'-diphosphate, disodium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Adenosine 5'-diphosphate, disodium salt hydrate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.