missing translation for 'onlineSavingsMsg'
Learn More
Adenosine 5'-monophosphate, 99%, Thermo Scientific Chemicals Product Code.: 10274600

Adenosine 5'-monophosphate, 99%, Thermo Scientific Chemicals

Product Code. 10274600
25 g, Glass bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10274600

Brand: Thermo Scientific Acros 102790250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-19-8
Molecular Formula C10H14N5O7P
Molecular Weight (g/mol) 347.22
MDL Number MFCD00005750
InChI Key UDMBCSSLTHHNCD-YPLCUDRINA-N
Synonym adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentasideShow MoreShow Less
PubChem CID 6083
ChEBI CHEBI:16027
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Absorbance 0.76 to 0.80 (A250/A260), 0.14 to 0.16 (A280/A260)
CAS Min % 98.5
CAS Max % 100.0
Melting Point 178.0°C to 185.0°C
Color Colorless to White
Loss on Drying 6% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 26, IV, 3615
Merck Index 15, 146
Solubility Information Solubility in water: soluble. Other solubilities: readily soluble in boiling water, soluble in ethanol
Spectrophotometry 98% min. (lambda max. 260nm,pH 7) molar absorptivity 15400+/-2%,on dry basis
Transmittance 95.0% min. (c=1, 0.2N NaOH, 430nm) 1cm cell
Formula Weight 347.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine 5′-monophosphate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467551 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number102791000,102790050,102790250
Lot NumberA0448620 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number102791000,102790050,102790250
Lot NumberA0403035 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number102791000,102790050,102790250
Lot NumberA0395274 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number102791000,102790050,102790250
Lot NumberA0417099 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number102791000,102790050,102790250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Adenosine 5'-monophosphate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.